Dendritic Cell Marker DCIR2 Monoclonal Antibody (33D1), eFluor™ 660, eBioscience™ - FAQs
Dendritic Cell Marker DCIR2 Monoclonal Antibody (33D1), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Dendritic Cell Marker DCIR2 Monoclonal Antibody (33D1), eFluor™ 660, eBioscience™ - FAQs (50-5884-80, 50-5884-82)