GL7 Monoclonal Antibody (GL-7 (GL7)), eFluor™ 660, eBioscience™ - FAQs
GL7 Monoclonal Antibody (GL-7 (GL7)), eFluor™ 660, eBioscience™ - FAQs
View additional product information for GL7 Monoclonal Antibody (GL-7 (GL7)), eFluor™ 660, eBioscience™ - FAQs (50-5902-80, 50-5902-82)