Themis Monoclonal Antibody (1TMYS), eFluor™ 660, eBioscience™ - FAQs
Themis Monoclonal Antibody (1TMYS), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Themis Monoclonal Antibody (1TMYS), eFluor™ 660, eBioscience™ - FAQs (50-5918-82)