AHR Monoclonal Antibody (4MEJJ), eFluor™ 660, eBioscience™ - FAQs
AHR Monoclonal Antibody (4MEJJ), eFluor™ 660, eBioscience™ - FAQs
View additional product information for AHR Monoclonal Antibody (4MEJJ), eFluor™ 660, eBioscience™ - FAQs (50-5925-80, 50-5925-82)