AIRE Monoclonal Antibody (5H12), eFluor™ 660, eBioscience™ - FAQs
AIRE Monoclonal Antibody (5H12), eFluor™ 660, eBioscience™ - FAQs
View additional product information for AIRE Monoclonal Antibody (5H12), eFluor™ 660, eBioscience™ - FAQs (50-5934-82, 50-5934-80)