CD275 (B7-H2) Monoclonal Antibody (HK5.3), eFluor™ 660, eBioscience™ - FAQs
CD275 (B7-H2) Monoclonal Antibody (HK5.3), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD275 (B7-H2) Monoclonal Antibody (HK5.3), eFluor™ 660, eBioscience™ - FAQs (50-5985-80, 50-5985-82)