c-Rel Monoclonal Antibody (1RELAH5), eFluor™ 660, eBioscience™ - FAQs
c-Rel Monoclonal Antibody (1RELAH5), eFluor™ 660, eBioscience™ - FAQs
View additional product information for c-Rel Monoclonal Antibody (1RELAH5), eFluor™ 660, eBioscience™ - FAQs (50-6111-80, 50-6111-82)