Smooth Muscle Myosin Monoclonal Antibody (SMMS-1), eFluor™ 660, eBioscience™ - FAQs
Smooth Muscle Myosin Monoclonal Antibody (SMMS-1), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Smooth Muscle Myosin Monoclonal Antibody (SMMS-1), eFluor™ 660, eBioscience™ - FAQs (50-6400-80)