GFP Monoclonal Antibody (5F12.4), eFluor™ 660, eBioscience™ - FAQs
GFP Monoclonal Antibody (5F12.4), eFluor™ 660, eBioscience™ - FAQs
View additional product information for GFP Monoclonal Antibody (5F12.4), eFluor™ 660, eBioscience™ - FAQs (50-6498-80, 50-6498-82)