TOX Monoclonal Antibody (TXRX10), eFluor™ 660, eBioscience™ - FAQs
TOX Monoclonal Antibody (TXRX10), eFluor™ 660, eBioscience™ - FAQs
View additional product information for TOX Monoclonal Antibody (TXRX10), eFluor™ 660, eBioscience™ - FAQs (50-6502-80, 50-6502-82)