Myosin 4 Monoclonal Antibody (MF20), eFluor™ 660, eBioscience™ - FAQs
Myosin 4 Monoclonal Antibody (MF20), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Myosin 4 Monoclonal Antibody (MF20), eFluor™ 660, eBioscience™ - FAQs (50-6503-82, 50-6503-80)