Oligodendrocyte Marker O1 Monoclonal Antibody (O1), eFluor™ 660, eBioscience™ - FAQs
Oligodendrocyte Marker O1 Monoclonal Antibody (O1), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Oligodendrocyte Marker O1 Monoclonal Antibody (O1), eFluor™ 660, eBioscience™ - FAQs (50-6506-80, 50-6506-82)