Synaptophysin Monoclonal Antibody (EP10), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Synaptophysin Monoclonal Antibody (EP10), eFluor™ 660, eBioscience™ - FAQs (50-6525-80, 50-6525-82)