Bcl-2 Monoclonal Antibody (10C4), eFluor™ 660, eBioscience™ - FAQs
Bcl-2 Monoclonal Antibody (10C4), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Bcl-2 Monoclonal Antibody (10C4), eFluor™ 660, eBioscience™ - FAQs (50-6992-42, 50-6992-41)