IL-23 p19 Monoclonal Antibody (fc23cpg), eFluor™ 660, eBioscience™ - FAQs
IL-23 p19 Monoclonal Antibody (fc23cpg), eFluor™ 660, eBioscience™ - FAQs
View additional product information for IL-23 p19 Monoclonal Antibody (fc23cpg), eFluor™ 660, eBioscience™ - FAQs (50-7023-80, 50-7023-82)