IL-4 Monoclonal Antibody (OX81), eFluor™ 660, eBioscience™ - FAQs
IL-4 Monoclonal Antibody (OX81), eFluor™ 660, eBioscience™ - FAQs
View additional product information for IL-4 Monoclonal Antibody (OX81), eFluor™ 660, eBioscience™ - FAQs (50-7045-82, 50-7045-80)