IL-9 Monoclonal Antibody (MH9A4), eFluor™ 660, eBioscience™ - FAQs
IL-9 Monoclonal Antibody (MH9A4), eFluor™ 660, eBioscience™ - FAQs
View additional product information for IL-9 Monoclonal Antibody (MH9A4), eFluor™ 660, eBioscience™ - FAQs (50-7097-42, 50-7097-41)