IL-10 Monoclonal Antibody (JES3-9D7), eFluor™ 660, eBioscience™ - FAQs
IL-10 Monoclonal Antibody (JES3-9D7), eFluor™ 660, eBioscience™ - FAQs
View additional product information for IL-10 Monoclonal Antibody (JES3-9D7), eFluor™ 660, eBioscience™ - FAQs (50-7108-41, 50-7108-42)