IL-12/IL-23 p40 Monoclonal Antibody (C17.8), eFluor™ 660, eBioscience™ - FAQs
IL-12/IL-23 p40 Monoclonal Antibody (C17.8), eFluor™ 660, eBioscience™ - FAQs
View additional product information for IL-12/IL-23 p40 Monoclonal Antibody (C17.8), eFluor™ 660, eBioscience™ - FAQs (50-7123-82, 50-7123-80)