IL-12/IL-23 p40 Monoclonal Antibody (C8.6), eFluor™ 660, eBioscience™ - FAQs
View additional product information for IL-12/IL-23 p40 Monoclonal Antibody (C8.6), eFluor™ 660, eBioscience™ - FAQs (50-7129-42, 50-7129-41)