IL-13 Monoclonal Antibody (eBio13A), eFluor™ 660, eBioscience™ - FAQs
IL-13 Monoclonal Antibody (eBio13A), eFluor™ 660, eBioscience™ - FAQs
View additional product information for IL-13 Monoclonal Antibody (eBio13A), eFluor™ 660, eBioscience™ - FAQs (50-7133-80, 50-7133-82)