IL-17F Monoclonal Antibody (SHLR17), eFluor™ 660, eBioscience™ - FAQs
IL-17F Monoclonal Antibody (SHLR17), eFluor™ 660, eBioscience™ - FAQs
View additional product information for IL-17F Monoclonal Antibody (SHLR17), eFluor™ 660, eBioscience™ - FAQs (50-7169-42)