IL-17A Monoclonal Antibody (eBio64CAP17), eFluor™ 660, eBioscience™ - FAQs
IL-17A Monoclonal Antibody (eBio64CAP17), eFluor™ 660, eBioscience™ - FAQs
View additional product information for IL-17A Monoclonal Antibody (eBio64CAP17), eFluor™ 660, eBioscience™ - FAQs (50-7178-42, 50-7178-41)