CD196 (CCR6) Monoclonal Antibody (sirx6), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD196 (CCR6) Monoclonal Antibody (sirx6), eFluor™ 660, eBioscience™ - FAQs (50-7196-82, 50-7196-80)