IL-21 Monoclonal Antibody (mhalx21), eFluor™ 660, eBioscience™ - FAQs
IL-21 Monoclonal Antibody (mhalx21), eFluor™ 660, eBioscience™ - FAQs
View additional product information for IL-21 Monoclonal Antibody (mhalx21), eFluor™ 660, eBioscience™ - FAQs (50-7213-82)