IL-22 Monoclonal Antibody (22URTI), eFluor™ 660, eBioscience™ - FAQs
View additional product information for IL-22 Monoclonal Antibody (22URTI), eFluor™ 660, eBioscience™ - FAQs (50-7229-41, 50-7229-42)