IFN gamma Monoclonal Antibody (DB-1 (DB1)), eFluor™ 660, eBioscience™ - FAQs
IFN gamma Monoclonal Antibody (DB-1 (DB1)), eFluor™ 660, eBioscience™ - FAQs
View additional product information for IFN gamma Monoclonal Antibody (DB-1 (DB1)), eFluor™ 660, eBioscience™ - FAQs (50-7310-80)