IFN gamma Monoclonal Antibody (XMG1.2), eFluor™ 660, eBioscience™ - FAQs
IFN gamma Monoclonal Antibody (XMG1.2), eFluor™ 660, eBioscience™ - FAQs
View additional product information for IFN gamma Monoclonal Antibody (XMG1.2), eFluor™ 660, eBioscience™ - FAQs (50-7311-80, 50-7311-82)