IFN gamma Monoclonal Antibody (4S.B3), eFluor™ 660, eBioscience™ - FAQs
IFN gamma Monoclonal Antibody (4S.B3), eFluor™ 660, eBioscience™ - FAQs
View additional product information for IFN gamma Monoclonal Antibody (4S.B3), eFluor™ 660, eBioscience™ - FAQs (50-7319-41)