G-CSF Monoclonal Antibody (8F5CSF), eFluor™ 660, eBioscience™ - FAQs
G-CSF Monoclonal Antibody (8F5CSF), eFluor™ 660, eBioscience™ - FAQs
View additional product information for G-CSF Monoclonal Antibody (8F5CSF), eFluor™ 660, eBioscience™ - FAQs (50-7351-42)