G-CSF Monoclonal Antibody (9B4CSF), eFluor™ 660, eBioscience™ - FAQs
G-CSF Monoclonal Antibody (9B4CSF), eFluor™ 660, eBioscience™ - FAQs
View additional product information for G-CSF Monoclonal Antibody (9B4CSF), eFluor™ 660, eBioscience™ - FAQs (50-7353-82, 50-7353-80)