IL-12 p35 Monoclonal Antibody (SNKY35), eFluor™ 660, eBioscience™ - FAQs
IL-12 p35 Monoclonal Antibody (SNKY35), eFluor™ 660, eBioscience™ - FAQs
View additional product information for IL-12 p35 Monoclonal Antibody (SNKY35), eFluor™ 660, eBioscience™ - FAQs (50-7359-42)