IL-38 Monoclonal Antibody (H127C), eFluor™ 660, eBioscience™ - FAQs
IL-38 Monoclonal Antibody (H127C), eFluor™ 660, eBioscience™ - FAQs
View additional product information for IL-38 Monoclonal Antibody (H127C), eFluor™ 660, eBioscience™ - FAQs (50-7385-80)