IL-17F Monoclonal Antibody (eBio18F10), eFluor™ 660, eBioscience™ - FAQs
IL-17F Monoclonal Antibody (eBio18F10), eFluor™ 660, eBioscience™ - FAQs
View additional product information for IL-17F Monoclonal Antibody (eBio18F10), eFluor™ 660, eBioscience™ - FAQs (50-7471-82, 50-7471-80)