IL-23 p19 Monoclonal Antibody (23dcdp), eFluor™ 660, eBioscience™ - FAQs
IL-23 p19 Monoclonal Antibody (23dcdp), eFluor™ 660, eBioscience™ - FAQs
View additional product information for IL-23 p19 Monoclonal Antibody (23dcdp), eFluor™ 660, eBioscience™ - FAQs (50-7823-41, 50-7823-42)