IL-9 Monoclonal Antibody (RM9A4), eFluor™ 660, eBioscience™ - FAQs
IL-9 Monoclonal Antibody (RM9A4), eFluor™ 660, eBioscience™ - FAQs
View additional product information for IL-9 Monoclonal Antibody (RM9A4), eFluor™ 660, eBioscience™ - FAQs (50-8091-82, 50-8091-80)