IL-27 p28 Monoclonal Antibody (3D1p28), eFluor™ 660, eBioscience™ - FAQs
IL-27 p28 Monoclonal Antibody (3D1p28), eFluor™ 660, eBioscience™ - FAQs
View additional product information for IL-27 p28 Monoclonal Antibody (3D1p28), eFluor™ 660, eBioscience™ - FAQs (50-8277-41, 50-8277-42)