Granzyme K Monoclonal Antibody (G3H69), eFluor™ 660, eBioscience™ - FAQs
Granzyme K Monoclonal Antibody (G3H69), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Granzyme K Monoclonal Antibody (G3H69), eFluor™ 660, eBioscience™ - FAQs (50-8897-42, 50-8897-41)