Granzyme B Monoclonal Antibody (NGZB), eFluor™ 660, eBioscience™ - FAQs
Granzyme B Monoclonal Antibody (NGZB), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Granzyme B Monoclonal Antibody (NGZB), eFluor™ 660, eBioscience™ - FAQs (50-8898-82, 50-8898-80)