Pan Cytokeratin Monoclonal Antibody (AE1/AE3), eFluor™ 660, eBioscience™ - FAQs
Pan Cytokeratin Monoclonal Antibody (AE1/AE3), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Pan Cytokeratin Monoclonal Antibody (AE1/AE3), eFluor™ 660, eBioscience™ - FAQs (50-9003-82, 50-9003-80)