CD368 (Clec6) Monoclonal Antibody (9B9), eFluor™ 660, eBioscience™ - FAQs
CD368 (Clec6) Monoclonal Antibody (9B9), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD368 (Clec6) Monoclonal Antibody (9B9), eFluor™ 660, eBioscience™ - FAQs (50-9113-42, 50-9113-41)