HSP70 Monoclonal Antibody (1B2hsp70), eFluor™ 660, eBioscience™ - FAQs
HSP70 Monoclonal Antibody (1B2hsp70), eFluor™ 660, eBioscience™ - FAQs
View additional product information for HSP70 Monoclonal Antibody (1B2hsp70), eFluor™ 660, eBioscience™ - FAQs (50-9115-80)