Galectin 9 Monoclonal Antibody (9M1-3), eFluor™ 660, eBioscience™ - FAQs
Galectin 9 Monoclonal Antibody (9M1-3), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Galectin 9 Monoclonal Antibody (9M1-3), eFluor™ 660, eBioscience™ - FAQs (50-9116-41, 50-9116-42)