Survivin Monoclonal Antibody (STLALYV), eFluor™ 660, eBioscience™ - FAQs
Survivin Monoclonal Antibody (STLALYV), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Survivin Monoclonal Antibody (STLALYV), eFluor™ 660, eBioscience™ - FAQs (50-9176-42)