Granzyme A Monoclonal Antibody (CB9), eFluor™ 660, eBioscience™ - FAQs
Granzyme A Monoclonal Antibody (CB9), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Granzyme A Monoclonal Antibody (CB9), eFluor™ 660, eBioscience™ - FAQs (50-9177-42)