IL-17FF (homodimer) Monoclonal Antibody (WU24A4P), eFluor™ 660, eBioscience™ - FAQs
IL-17FF (homodimer) Monoclonal Antibody (WU24A4P), eFluor™ 660, eBioscience™ - FAQs
View additional product information for IL-17FF (homodimer) Monoclonal Antibody (WU24A4P), eFluor™ 660, eBioscience™ - FAQs (50-9178-42)