IL-31 Monoclonal Antibody (31SNEZE), eFluor™ 660, eBioscience™ - FAQs
IL-31 Monoclonal Antibody (31SNEZE), eFluor™ 660, eBioscience™ - FAQs
View additional product information for IL-31 Monoclonal Antibody (31SNEZE), eFluor™ 660, eBioscience™ - FAQs (50-9319-42)