CD326 (EpCAM) Monoclonal Antibody (1B7), eFluor™ 660, eBioscience™ - FAQs
CD326 (EpCAM) Monoclonal Antibody (1B7), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD326 (EpCAM) Monoclonal Antibody (1B7), eFluor™ 660, eBioscience™ - FAQs (50-9326-41, 50-9326-42)