CD45RB Monoclonal Antibody (PD7/26), eFluor™ 660, eBioscience™ - FAQs
CD45RB Monoclonal Antibody (PD7/26), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD45RB Monoclonal Antibody (PD7/26), eFluor™ 660, eBioscience™ - FAQs (50-9458-82)