IDO Monoclonal Antibody (mIDO-48), eFluor™ 660, eBioscience™ - FAQs
IDO Monoclonal Antibody (mIDO-48), eFluor™ 660, eBioscience™ - FAQs
View additional product information for IDO Monoclonal Antibody (mIDO-48), eFluor™ 660, eBioscience™ - FAQs (50-9473-80, 50-9473-82)